The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164798.1 | NP_001158270.1 | 7 - 8 | 1000 | 80 |
NM_001164799.1 | NP_001158271.1 | 7 - 8 | 1000 | 80 | |
NM_001164800.1 | NP_001158272.1 | 7 - 8 | 1000 | 80 | |
NM_001164801.1 | NP_001158273.1 | 7 - 8 | 1000 | 80 | |
NM_001164802.1 | NP_001158274.1 | 7 - 8 | 1000 | 80 | |
NM_024249.5 | NP_077211.4 | 7 - 8 | 1000 | 80 | |
GenBank mRNA | AK007977.1 | - | 7 - 8 | 890 | 80 |
AK032245.1 | - | 7 - 8 | 924 | 80 | |
AK090050.1 | - | 7 - 8 | 976 | 80 | |
AK159948.1 | - | 7 - 8 | 962 | 80 | |
AK165672.1 | - | 7 - 8 | 965 | 80 | |
BC082300.1 | - | 7 - 8 | 971 | 80 | |
BC088811.1 | - | 7 - 8 | 936 | 80 |