The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199283.1 | NP_001186212.1 | 4 - 5 | 766 | 73 |
NM_001199284.1 | NP_001186213.1 | 4 - 5 | 548 | 73 | |
NM_173388.2 | NP_775564.1 | 4 - 5 | 511 | 73 | |
GenBank mRNA | AB120363.1 | - | 4 - 5 | 544 | 73 |
AK143773.1 | - | 4 - 5 | 609 | 73 | |
AK159144.1 | - | 4 - 5 | 734 | 73 | |
AK171104.1 | - | 4 - 5 | 766 | 73 | |
AK172685.1 | - | 4 - 5 | 536 | 73 | |
BC042513.1 | - | 4 - 5 | 507 | 73 |