The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035123.3 | NP_001030295.1 | 6 - 7 | 1133 | 66 |
GenBank mRNA | AK010304.1 | - | 6 - 7 | 1134 | 66 |
AK075597.1 | - | 6 - 7 | 1136 | 66 | |
AK167753.1 | - | 6 - 7 | 1115 | 66 | |
AK181998.1 | - | 2 - 3 | 318 | 66 | |
BC139198.1 | - | 6 - 7 | 1165 | 66 | |
BC139199.1 | - | 6 - 7 | 1165 | 66 |