Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyld
Entrez Gene ID
74256
Gene Name
cylindromatosis (turban tumor syndrome)
Gene Aliases
2010013M14Rik, 2900009M21Rik, C130039D01Rik, CDMT, CYLD1, EAC, mKIAA0849
Chromosome Location
Chr.8: 88697001 - 88751946 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128170.2 NP_001121642.1 17 - 18 2901 72
NM_001128171.2 NP_001121643.1 17 - 18 3026 72
NM_001276279.1 NP_001263208.1 15 - 16 2477 72
NM_173369.3 NP_775545.1 16 - 17 2892 72
GenBank mRNA AK122389.1 - 17 - 18 3007 72
AK133479.1 - 16 - 17 2856 72
AK134926.1 - 4 - 5 584 72
AK167424.1 - 15 - 16 2441 72
BC023702.1 - 1 - 2 150 72
BC024596.1 - 4 - 5 570 72
BC028885.1 - 9 - 10 2547 72
BC042438.1 - 16 - 17 2845 72
BC049879.1 - 7 - 8 754 72

View Full Product Details