The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146217.4 | NP_666329.2 | 2 - 3 | 589 | 77 |
GenBank mRNA | AK036738.1 | - | 2 - 3 | 198 | 77 |
AK044451.1 | - | 2 - 3 | 340 | 77 | |
AK085725.1 | - | 2 - 3 | 530 | 77 | |
AK150227.1 | - | 2 - 3 | 517 | 77 | |
AK158139.1 | - | 2 - 3 | 538 | 77 | |
AY223875.1 | - | 2 - 3 | 529 | 77 | |
BC033273.1 | - | - | 464 | 77 | |
BM943271.1 | - | 2 - 3 | 234 | 77 |