The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122594.2 | NP_001116066.2 | 12 - 13 | 1891 | 66 |
GenBank mRNA | AK048260.1 | - | 12 - 13 | 1848 | 66 |
BC117961.1 | - | 12 - 13 | 1891 | 66 | |
BC117962.1 | - | 12 - 13 | 1883 | 66 |