Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phlpp2
Entrez Gene ID
244650
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
AI481772, C130044A18Rik, Phlppl
Chromosome Location
Chr.8: 109868519 - 109944671 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122594.2 NP_001116066.2 18 - 19 2928 66
GenBank mRNA AK048260.1 - 18 - 19 2885 66
BC117961.1 - 18 - 19 2928 66
BC117962.1 - 18 - 19 2920 66

View Full Product Details