The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145831.1 | NP_001139303.1 | 10 - 11 | 1406 | 94 |
NM_001145832.1 | NP_001139304.1 | 6 - 7 | 890 | 94 | |
NM_010631.2 | NP_034761.3 | 8 - 9 | 1233 | 94 | |
GenBank mRNA | AF013118.2 | - | 4 - 5 | 869 | 94 |
AK132895.1 | - | 6 - 7 | 890 | 94 | |
AK135926.1 | - | 3 - 4 | 432 | 94 | |
AK166482.1 | - | 10 - 11 | 1385 | 94 | |
BC016118.1 | - | 3 - 4 | 437 | 94 | |
BC070429.1 | - | 8 - 9 | 1233 | 94 | |
D49546.1 | - | 8 - 9 | 1248 | 94 |