The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029049.1 | NP_083325.1 | 3 - 4 | 1310 | 89 |
GenBank mRNA | AB235902.1 | - | 3 - 4 | 1225 | 89 |
AK015440.1 | - | 3 - 4 | 1269 | 89 | |
BC145091.1 | - | 3 - 4 | 1310 | 89 | |
BC171943.1 | - | 3 - 4 | 1310 | 89 |