The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197944.1 | NP_922935.1 | 2 - 3 | 167 | 97 |
GenBank mRNA | AK157000.1 | - | - | 200 | 97 |
AY319653.1 | - | 2 - 3 | 167 | 97 | |
BC094291.1 | - | 1 - 2 | 143 | 97 |