Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp2b1
Entrez Gene ID
67972
Gene Name
ATPase, Ca++ transporting, plasma membrane 1
Gene Aliases
2810442I22Rik, E130111D10Rik, Pmca1
Chromosome Location
Chr.10: 98914359 - 99026143 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026482.2 NP_080758.1 16 - 17 3078 82
GenBank mRNA BC029045.1 - 10 - 11 1718 82
BC030901.1 - 10 - 11 1719 82
BC049262.1 - 10 - 11 1718 82
BC057180.1 - 1 - 2 93 82

View Full Product Details