Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rundc3a
Entrez Gene ID
51799
Gene Name
RUN domain containing 3A
Gene Aliases
RPIP-8, Rap2ip, Rpip8
Chromosome Location
Chr.11: 102393403 - 102402939 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252347.1 NP_001239276.1 6 - 7 906 75
NM_016759.4 NP_058039.1 6 - 7 906 75
GenBank mRNA AK002856.1 - 2 - 3 204 75
BC022745.1 - 4 - 5 631 75
BC046319.1 - 6 - 7 806 75
U73941.1 - 6 - 7 633 75

View Full Product Details