Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pecam1
- Entrez Gene ID
-
18613
- Gene Name
- platelet/endothelial cell adhesion molecule 1
- Gene Aliases
-
C85791, Cd31, PECAM-1, Pecam
- Chromosome Location
-
Chr.11: 106654217 - 106750623 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK084527.1 |
- |
14 - 15 |
2338 |
82 |
View Full Product Details