Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nol11
Entrez Gene ID
68979
Gene Name
nucleolar protein 11
Gene Aliases
1500002M01Rik, AU015220, AU019874
Chromosome Location
Chr.11: 107166660 - 107189381 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161329.1 NP_001154801.1 7 - 8 978 82
NM_133702.2 NP_598463.2 7 - 8 978 82
GenBank mRNA AK005123.1 - 7 - 8 867 82
AK038696.1 - 7 - 8 869 82
AK076704.1 - 7 - 8 979 82
AK078524.1 - 7 - 8 869 82
AK157743.1 - 7 - 8 918 82
BC004785.1 - 2 - 3 257 82
BC024874.1 - 7 - 8 857 82

View Full Product Details