The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001445.2 | NP_001001445.1 | 12 - 13 | 2041 | 76 |
GenBank mRNA | AB180097.1 | - | 11 - 12 | 1789 | 76 |
AJ620495.1 | - | 11 - 12 | 1829 | 76 | |
AK051315.1 | - | 12 - 13 | 1899 | 76 | |
AY445519.1 | - | 11 - 12 | 1789 | 76 | |
AY452083.1 | - | 11 - 12 | 1822 | 76 | |
AY452084.1 | - | 11 - 12 | 1792 | 76 |