Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ict1
- Entrez Gene ID
-
68572
- Gene Name
- immature colon carcinoma transcript 1
- Gene Aliases
-
1110001A02Rik, 1110002E03Rik, MRP-L58
- Chromosome Location
-
Chr.11: 115403725 - 115410919 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC079614.1 |
- |
- |
246 |
93 |
View Full Product Details