Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pcyt2
- Entrez Gene ID
-
68671
- Gene Name
- phosphate cytidylyltransferase 2, ethanolamine
- Gene Aliases
-
1110033E03Rik, ET
- Chromosome Location
-
Chr.11: 120610086 - 120617949 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK166319.1 |
- |
6 - 7 |
604 |
96 |
View Full Product Details