Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Map2k6
- Entrez Gene ID
-
26399
- Gene Name
- mitogen-activated protein kinase kinase 6
- Gene Aliases
-
MEK6, MKK6, Prkmk6, SAPKK3
- Chromosome Location
-
Chr.11: 110399102 - 110516212 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X98067.1 |
- |
1 - 2 |
391 |
61 |
View Full Product Details