Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasd2
Entrez Gene ID
75141
Gene Name
RASD family, member 2
Gene Aliases
4930526B11Rik, AU045414, Rhes, TEM-2, TEM2
Chromosome Location
Chr.8: 75213944 - 75224113 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029182.1 NP_083458.1 2 - 3 419 52
GenBank mRNA AK015898.1 - 2 - 3 650 52
BC036988.1 - 2 - 3 430 52

View Full Product Details