The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163791.1 | NP_001157263.1 | 1 - 2 | 354 | 121 |
GenBank mRNA | AK078405.1 | - | 1 - 2 | 88 | 121 |
AK163256.1 | - | 1 - 2 | 298 | 121 |