The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291185.1 | NP_001278114.1 | 8 - 9 | 1100 | 67 |
NM_011497.4 | NP_035627.1 | 8 - 9 | 1178 | 67 | |
GenBank mRNA | AF007817.1 | - | 8 - 9 | 1125 | 67 |
AK077513.1 | - | 8 - 9 | 1056 | 67 | |
AK085861.1 | - | 8 - 9 | 1052 | 67 | |
AK145968.1 | - | 8 - 9 | 1066 | 67 | |
AK169363.1 | - | 8 - 9 | 1055 | 67 | |
AY336976.1 | - | 7 - 8 | 1002 | 67 | |
BC005425.1 | - | 8 - 9 | 1045 | 67 | |
U69106.1 | - | 8 - 9 | 1036 | 67 | |
U80932.1 | - | 8 - 9 | 1045 | 67 |