The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024927.2 | NP_001020098.1 | 10 - 11 | 1469 | 59 |
NM_001081641.1 | NP_001075110.1 | 9 - 10 | 1421 | 59 | |
GenBank mRNA | AK147444.1 | - | 10 - 11 | 1426 | 59 |
AK147630.1 | - | 9 - 10 | 1254 | 59 | |
BC112411.1 | - | 9 - 10 | 1844 | 59 |