Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pitpnm3
Entrez Gene ID
327958
Gene Name
PITPNM family member 3
Gene Aliases
A330068P14Rik, AI848332, Ackr6, Gm880
Chromosome Location
Chr.11: 72047528 - 72135889 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024927.2 NP_001020098.1 7 - 8 988 61
NM_001081641.1 NP_001075110.1 6 - 7 940 61
GenBank mRNA AK039594.1 - 6 - 7 1181 61
AK147444.1 - 7 - 8 945 61
AK147630.1 - 6 - 7 773 61
BC112411.1 - 6 - 7 1363 61

View Full Product Details