The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026810.2 | NP_081086.2 | 2 - 3 | 293 | 120 |
GenBank mRNA | AF250844.1 | - | 2 - 3 | 209 | 120 |
AK038265.1 | - | 2 - 3 | 286 | 120 | |
AK145646.1 | - | 2 - 3 | 239 | 120 | |
AK163861.1 | - | 2 - 3 | 227 | 120 | |
AK168849.1 | - | 2 - 3 | 232 | 120 | |
BC021815.1 | - | 2 - 3 | 247 | 120 |