Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mlh1
Entrez Gene ID
17350
Gene Name
mutL homolog 1 (E. coli)
Gene Aliases
1110035C23Rik, AI317206, AI325952, AI561766
Chromosome Location
Chr.9: 111228228 - 111271786 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026810.2 NP_081086.2 7 - 8 673 96
GenBank mRNA AF250844.1 - 7 - 8 589 96
AK038265.1 - 7 - 8 666 96
AK145646.1 - 7 - 8 620 96
AK163861.1 - 7 - 8 608 96
AK168849.1 - 7 - 8 612 96
BC021815.1 - 7 - 8 627 96

View Full Product Details