Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Marveld3
Entrez Gene ID
73608
Gene Name
MARVEL (membrane-associating) domain containing 3
Gene Aliases
1810006A16Rik, AI642133, Marvd3, Mrvldc3
Chromosome Location
Chr.8: 109947909 - 109962205 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212447.2 NP_997612.2 2 - 3 619 88
GenBank mRNA BC025851.1 - 2 - 3 582 88

View Full Product Details