Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prpsap1
Entrez Gene ID
67763
Gene Name
phosphoribosyl pyrophosphate synthetase-associated protein 1
Gene Aliases
5730409F23Rik, PAP39
Chromosome Location
Chr.11: 116470816 - 116494652 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026364.1 NP_080640.1 1 - 2 304 93
GenBank mRNA AK011290.1 - 1 - 2 274 93
AK077439.1 - 1 - 2 307 93
AK082484.1 - 1 - 2 273 93
AK160763.1 - 1 - 2 288 93
AK218181.1 - 1 - 2 38 93
BC029621.1 - 1 - 2 295 93
BY261597.1 - 1 - 2 306 93

View Full Product Details