Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp36
Entrez Gene ID
72344
Gene Name
ubiquitin specific peptidase 36
Gene Aliases
2700002L06Rik, mKIAA1453
Chromosome Location
Chr.11: 118259651 - 118290244 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033528.1 NP_001028700.1 6 - 7 1093 73
GenBank mRNA AK028572.1 - 6 - 7 1138 73
AK049205.1 - 6 - 7 923 73
AK129363.1 - 6 - 7 891 73

View Full Product Details