The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133660.3 | NP_598421.1 | 9 - 10 | 1277 | 67 |
GenBank mRNA | AK144549.1 | - | 9 - 10 | 1262 | 67 |
BC019208.1 | - | 9 - 10 | 1208 | 67 |