The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164677.1 | NP_001158149.1 | 1 - 2 | 360 | 114 |
NM_001164678.1 | NP_001158150.1 | 1 - 2 | 360 | 114 | |
NM_011052.2 | NP_035182.2 | 1 - 2 | 360 | 114 | |
GenBank mRNA | AF119955.1 | - | 1 - 2 | 321 | 114 |
AJ005073.1 | - | 1 - 2 | 208 | 114 | |
AJ005074.1 | - | 1 - 2 | 208 | 114 | |
AK029827.1 | - | 1 - 2 | 327 | 114 | |
AK031256.1 | - | 1 - 2 | 346 | 114 | |
AK033943.1 | - | 1 - 2 | 348 | 115 | |
AK129340.1 | - | 1 - 2 | 330 | 114 | |
AK145893.1 | - | 1 - 2 | 347 | 114 | |
AK147736.1 | - | 1 - 2 | 333 | 114 | |
AK167574.1 | - | 1 - 2 | 318 | 114 | |
AK169704.1 | - | 1 - 2 | 361 | 114 | |
BC026823.1 | - | 1 - 2 | 302 | 114 | |
BC051123.1 | - | 1 - 2 | 211 | 114 |