The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170684.2 | NP_733785.1 | 14 - 15 | 1664 | 90 |
GenBank mRNA | AI550750.1 | - | 1 - 2 | 77 | 90 |
AK049823.1 | - | 11 - 12 | 2806 | 90 | |
AK162900.1 | - | 13 - 14 | 2283 | 90 | |
BC119328.1 | - | 14 - 15 | 1563 | 90 | |
BC137595.1 | - | 14 - 15 | 1563 | 90 |