The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033371.3 | NP_001028543.1 | 5 - 6 | 887 | 101 |
NM_001170788.1 | NP_001164259.1 | 8 - 9 | 1216 | 101 | |
NM_001170789.1 | NP_001164260.1 | 5 - 6 | 893 | 101 | |
GenBank mRNA | AK133042.1 | - | 5 - 6 | 887 | 101 |
BC036564.1 | - | 3 - 4 | 515 | 101 | |
BC150957.1 | - | 5 - 6 | 879 | 101 | |
BC172120.1 | - | 5 - 6 | 885 | 101 |