Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rltpr
- Entrez Gene ID
-
234695
- Gene Name
- RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing
- Gene Aliases
-
CG1399-PB, D130029J02Rik, Gm585
- Chromosome Location
-
Chr.8: 105686010 - 105698195 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
HF678090.1 |
- |
7 - 8 |
532 |
80 |
View Full Product Details