Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfs1
Entrez Gene ID
18041
Gene Name
nitrogen fixation gene 1 (S. cerevisiae)
Gene Aliases
AA987187, m-Nfs1, m-Nfsl
Chromosome Location
Chr.2: 156123637 - 156144186 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010911.2 NP_035041.2 10 - 11 1190 76
GenBank mRNA AK005724.1 - 10 - 11 1180 76
AK075575.1 - 10 - 11 1188 76
AK132838.1 - 10 - 11 1191 76
AK148527.1 - 10 - 11 1191 76
AK159828.1 - 10 - 11 1179 76
AK189862.1 - 2 - 3 178 76
AK191681.1 - 2 - 3 178 76
AK193838.1 - 2 - 3 178 76
AK198789.1 - 2 - 3 178 76

View Full Product Details