Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il27ra
Entrez Gene ID
50931
Gene Name
interleukin 27 receptor, alpha
Gene Aliases
CRL1, IL-27R, Tccr, WSX-1, Wsx1, zcytor1
Chromosome Location
Chr.8: 84030286 - 84042629 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016671.3 NP_057880.2 11 - 12 1545 133
GenBank mRNA AF053005.1 - 11 - 12 1513 133
AF265243.1 - 11 - 12 1524 133
AK040457.1 - 9 - 10 1885 133
AK133753.1 - 11 - 12 1560 133
BC032878.1 - 11 - 12 1545 133

View Full Product Details