Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cirh1a
Entrez Gene ID
21771
Gene Name
cirrhosis, autosomal recessive 1A (human)
Gene Aliases
Cirhin, Naic, Teg-292, Tex292
Chromosome Location
Chr.8: 106893636 - 106923094 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011574.2 NP_035704.2 6 - 7 812 62
GenBank mRNA AK144455.1 - 6 - 7 797 62
AK146263.1 - 6 - 7 788 62
AK150862.1 - 6 - 7 787 62
AK168470.1 - 6 - 7 762 62
AK173318.1 - 5 - 6 753 62
AK178714.1 - 2 - 3 261 62
AK179441.1 - 1 - 2 44 62
AK180185.1 - 2 - 3 261 62
AK187526.1 - 2 - 3 261 62
AK195627.1 - 2 - 3 266 62
BC027399.1 - 2 - 3 311 62
BC068113.1 - 6 - 7 801 62

View Full Product Details