Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam92b
Entrez Gene ID
436062
Gene Name
family with sequence similarity 92, member B
Gene Aliases
1700120B06Rik
Chromosome Location
Chr.8: 120166397 - 120177486 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033980.2 NP_001029152.2 1 - 2 157 79
GenBank mRNA AK131790.1 - 1 - 2 157 79
BC147233.1 - 1 - 2 76 79
BC147234.1 - 1 - 2 76 79

View Full Product Details