The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271915.1 | NP_001258844.1 | 1 - 1 | 1362 | 75 |
NM_178879.4 | NP_849210.1 | 2 - 2 | 1174 | 75 | |
GenBank mRNA | AK032108.1 | - | 1 - 1 | 1363 | 75 |
AK053867.1 | - | 1 - 1 | 876 | 75 | |
AK169360.1 | - | 1 - 1 | 1358 | 75 | |
BC047209.1 | - | 1 - 1 | 1334 | 75 | |
BC108347.1 | - | 1 - 1 | 1312 | 75 | |
BC120583.1 | - | 1 - 1 | 1027 | 75 | |
BC120585.1 | - | 1 - 1 | 1027 | 75 |