The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026280.3 | NP_080556.1 | 3 - 4 | 466 | 75 |
GenBank mRNA | AK012601.1 | - | 3 - 4 | 438 | 75 |
AK075801.1 | - | 3 - 4 | 467 | 75 | |
BC147320.1 | - | 3 - 4 | 547 | 75 | |
BC147321.1 | - | 3 - 4 | 547 | 75 |