Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bpifa1
Entrez Gene ID
18843
Gene Name
BPI fold containing family A, member 1
Gene Aliases
LUNX, NASG, Plunc, SPLUNC1, SPURT
Chromosome Location
Chr.2: 154142880 - 154149217 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011126.3 NP_035256.2 2 - 3 298 129
GenBank mRNA BC054375.1 - 2 - 3 279 129
U69172.1 - 2 - 3 294 129

View Full Product Details