The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199015.1 | NP_001185944.1 | 3 - 4 | 304 | 77 |
NM_001199016.1 | NP_001185945.1 | 2 - 3 | 192 | 77 | |
NM_021291.3 | NP_067266.1 | 2 - 3 | 419 | 77 | |
GenBank mRNA | AA104782.1 | - | 2 - 3 | 253 | 77 |
AF192310.1 | - | 3 - 4 | 304 | 77 | |
AJ249198.1 | - | 2 - 3 | 419 | 77 | |
AK142541.1 | - | 2 - 3 | 193 | 77 | |
AK164714.1 | - | 2 - 3 | 191 | 77 | |
BC010746.1 | - | 3 - 4 | 282 | 77 |