The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153088.2 | NP_694728.1 | 1 - 2 | 354 | 85 |
GenBank mRNA | AK089478.1 | - | 1 - 2 | 261 | 85 |
AK167390.1 | - | 1 - 2 | 112 | 85 | |
AY028804.1 | - | 1 - 2 | 379 | 85 | |
BC049184.1 | - | 1 - 2 | 214 | 85 | |
BC058085.1 | - | 1 - 2 | 171 | 85 | |
BC065158.1 | - | 1 - 2 | 89 | 85 | |
BC079638.1 | - | 1 - 2 | 89 | 85 | |
BP765657.1 | - | 1 - 2 | 383 | 85 |