The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018796.3 | NP_061266.2 | 3 - 4 | 705 | 88 |
GenBank mRNA | AK027911.1 | - | 3 - 4 | 408 | 88 |
AK168191.1 | - | 3 - 4 | 407 | 88 | |
BC003899.1 | - | 3 - 4 | 401 | 88 | |
BC023139.1 | - | 3 - 4 | 417 | 88 | |
BC039635.1 | - | 3 - 4 | 420 | 88 |