The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045513.3 | NP_001038978.3 | 10 - 11 | 1621 | 97 |
GenBank mRNA | DV652147.1 | - | 2 - 3 | 162 | 97 |