Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nbeal1
Entrez Gene ID
269198
Gene Name
neurobeachin like 1
Gene Aliases
2310076G13Rik, A530050O19Rik, A530083I02Rik, AI662228, ALS2CR17
Chromosome Location
Chr.1: 60178629 - 60334705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173444.2 NP_775620.2 52 - 53 8008 111
GenBank mRNA AK041108.1 - 12 - 13 1346 111
BC055813.1 - 15 - 16 1737 111
BC060141.1 - 28 - 29 4139 111

View Full Product Details