Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sf3b1
Entrez Gene ID
81898
Gene Name
splicing factor 3b, subunit 1
Gene Aliases
155kDa, 2810001M05Rik, AA409119, Prp10, SAP155, SF3b155, TA-8, Targ4
Chromosome Location
Chr.1: 54985169 - 55027500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031179.2 NP_112456.2 17 - 18 2604 74
GenBank mRNA AB037890.1 - 17 - 18 2629 74
CN526934.1 - 2 - 3 211 74

View Full Product Details