Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stradb
Entrez Gene ID
227154
Gene Name
STE20-related kinase adaptor beta
Gene Aliases
AA792893, Als2cr2, B830008M19, D1Ucla2, ILPIP, ILPIPA, PRO1038, Papk, Syradb
Chromosome Location
Chr.1: 58973527 - 58995126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172656.5 NP_766244.4 2 - 3 387 145
GenBank mRNA AB057666.1 - 2 - 3 154 145
AB057667.1 - 2 - 3 154 145
AK035207.1 - 2 - 3 318 145
AK036678.1 - 2 - 3 382 145
AK046790.1 - - 360 145
AK080096.1 - 2 - 3 351 145
AK086974.1 - 2 - 3 318 145
AK140272.1 - 2 - 3 437 145
AK146079.1 - 2 - 3 200 145
BC037003.1 - 2 - 3 296 145
BC038492.1 - 2 - 3 386 145
BC055814.1 - 2 - 3 360 145
BC060956.1 - 2 - 3 244 145
CJ060534.1 - 2 - 3 437 145

View Full Product Details