The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172626.2 | NP_766214.2 | 7 - 8 | 1322 | 76 |
GenBank mRNA | AK042613.1 | - | 2 - 3 | 555 | 76 |
AK129330.1 | - | 1 - 2 | 166 | 76 | |
AY461716.1 | - | 7 - 8 | 1297 | 76 | |
BC054080.1 | - | 3 - 4 | 770 | 76 |