Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcgr1
Entrez Gene ID
14129
Gene Name
Fc receptor, IgG, high affinity I
Gene Aliases
AI323638, AV092959, CD64, FcgammaRI, IGGHAFC
Chromosome Location
Chr.3: 96282909 - 96293969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF143171.1 - 1 - 2 62 70
AF143172.1 - 1 - 2 62 70
AF143173.1 - 1 - 2 62 70
AF143174.1 - 1 - 2 62 70
AF143175.1 - 1 - 2 62 70
AF143176.1 - 1 - 2 62 70
AF143177.1 - 1 - 2 62 70
AF143178.1 - 1 - 2 62 70
AF143179.1 - 1 - 2 62 70

View Full Product Details