Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mysm1
Entrez Gene ID
320713
Gene Name
myb-like, SWIRM and MPN domains 1
Gene Aliases
C130067A03Rik, C530050H10Rik
Chromosome Location
Chr.4: 94942037 - 94979302 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177239.2 NP_796213.2 14 - 15 1779 74
GenBank mRNA AK173300.1 - 11 - 12 1521 74
BC150946.1 - 14 - 15 1775 74
BC151172.1 - 14 - 15 1775 74

View Full Product Details